5-Fluorobenzo[d]isoxazol-3(2H)-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC201389-250MG
PC201389-1G
APOSPC201389-250MGEA
522
EUR
APOSPC201389-250MG
APOSPC201389-1G
5-Fluorobenzo[d]isoxazol-3(2H)-one 97%
5-Fluorobenzo[d]isoxazol-3(2H)-one
Formel:
C₇H₄FNO₂ Molecular Weight: 153,111 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11846402 CAS-Nummer: 99822-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...