6-Fluorobenzo[d]isoxazol-3(2H)-one 95%

Lieferant: Apollo Scientific

PC201390-250MG PC201390-1G
APOSPC201390-250MGEA 144 EUR
APOSPC201390-250MG APOSPC201390-1G
6-Fluorobenzo[d]isoxazol-3(2H)-one 95%
6-Fluorobenzo[d]isoxazol-3(2H)-one
Formel: C₇H₄FNO₂
Molecular Weight: 153,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13180537
CAS-Nummer: 99822-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR