Amino(4-fluorophenyl)acetonitrile 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Cyano-4-fluorobenzylamine
PC2032-25G
APOSPC2032-25GEA
0
EUR
APOSPC2032-25G
Amino(4-fluorophenyl)acetonitrile 97%
Amino(4-fluorophenyl)acetonitrile
Formel:
C₈H₇FN₂ Molecular Weight: 150,153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236207 CAS-Nummer: 56464-70-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...