4,4,4-Trifluorobut-1-ene 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Trifluoromethyl)prop-1-ene
PC2062-5G
PC2062-25G
APOSPC2062-5GEA
719
EUR
APOSPC2062-5G
APOSPC2062-25G
4,4,4-Trifluorobut-1-ene 97%
4,4,4-Trifluorobut-1-ene
Formel:
C₄H₅F₃ Molecular Weight: 110,078 g/mol Siedepunkt: 11 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02262195 CAS-Nummer: 1524-26-1 UN: 3161 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...