4-Amino-3-bromo-2,5-difluorobenzonitrile

Lieferant: Apollo Scientific

PC210030-1G PC210030-500MG
APOSPC210030-1GEA 1070 EUR
APOSPC210030-1G APOSPC210030-500MG
4-Amino-3-bromo-2,5-difluorobenzonitrile
4-Amino-3-bromo-2,5-difluorobenzonitrile
Formel: C₇H₃BrF₂N₂
Molecular Weight: 233,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658642
CAS-Nummer: 112279-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR