N-Methyl-N'-[1-(2,4,6-trifluoro-phenyl)-methylidene]-hydrazine

Lieferant: Apollo Scientific

PC210038-500MG
APOSPC210038-500MGEA 214 EUR
APOSPC210038-500MG
N-Methyl-N'-[1-(2,4,6-trifluoro-phenyl)-methylidene]-hydrazine
N-Methyl-N'-[1-(2,4,6-trifluoro-phenyl)-methylidene]-hydrazine
Formel: C₈H₇F₃N₂
Molecular Weight: 188,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27942767
CAS-Nummer: 1264837-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR