4-Bromo-1-(4-fluorophenyl)butan-1-one

Lieferant: Apollo Scientific

PC210044-1G PC210044-500MG
APOSPC210044-1GEA 894 EUR
APOSPC210044-1G APOSPC210044-500MG
4-Bromo-1-(4-fluorophenyl)butan-1-one
4-Bromo-1-(4-fluorophenyl)butan-1-one
Formel: C₁₀H₁₀BrFO
Molecular Weight: 245,088 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16323231
CAS-Nummer: 40132-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR