4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-methyl-1H-pyrazol-5-amine

Lieferant: Apollo Scientific

PC210074-1G PC210074-500MG
APOSPC210074-1GEA 890 EUR
APOSPC210074-1G APOSPC210074-500MG
4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-methyl-1H-pyrazol-5-amine
4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-methyl-1H-pyrazol-5-amine
Formel: C₁₀H₈ClF₃N₄
Molecular Weight: 276.646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347881
CAS-Nummer: 1644602-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR