8-Chloro-3-(2,6-dichlorophenyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridine
Lieferant: Apollo Scientific
PC210081-1G
PC210081-500MG
APOSPC210081-1GEA
741
EUR
APOSPC210081-1G
APOSPC210081-500MG
8-Chloro-3-(2,6-dichlorophenyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridine
8-Chloro-3-(2,6-dichlorophenyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridine
Formel:
C₁₄H₆Cl₃F₃N₂ Molecular Weight: 365,565 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28506330 CAS-Nummer: 1708126-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...