4-Fluoro-5-iodo-1H-indazole ≥98%
Lieferant: Apollo Scientific
PC210132-1G
APOSPC210132-1GEA
119
EUR
APOSPC210132-1G
4-Fluoro-5-iodo-1H-indazole ≥98%
4-Fluoro-5-iodo-1H-indazole
Formel:
C₇H₄FIN₂ Molecular Weight: 262,023 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007929 CAS-Nummer: 1082041-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...