8-Fluoro-3,4-dihydronaphthalen-2(1H)-one ≥98%

Lieferant: Apollo Scientific

PC210135-100MG PC210135-1G
APOSPC210135-100MGEA 339 EUR
APOSPC210135-100MG APOSPC210135-1G
8-Fluoro-3,4-dihydronaphthalen-2(1H)-one ≥98%
8-Fluoro-3,4-dihydronaphthalen-2(1H)-one
Formel: C₁₀H₉FO
Molecular Weight: 164,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038616
CAS-Nummer: 127169-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR