4-Ethyl-3-fluorophenol ≥98%

Lieferant: Apollo Scientific

PC210136-1G PC210136-5G
APOSPC210136-1GEA 467 EUR
APOSPC210136-1G APOSPC210136-5G
4-Ethyl-3-fluorophenol ≥98%
4-Ethyl-3-fluorophenol
Formel: C₈H₉FO
Molecular Weight: 140,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658621
CAS-Nummer: 326493-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR