1-(2-(Difluoromethyl)phenyl)ethanol ≥98%

Lieferant: Apollo Scientific

PC210140-100MG PC210140-250MG
APOSPC210140-100MGEA 178 EUR
APOSPC210140-100MG APOSPC210140-250MG
1-(2-(Difluoromethyl)phenyl)ethanol ≥98%
1-(2-(Difluoromethyl)phenyl)ethanol
Formel: C₉H₁₀F₂O
Molecular Weight: 172,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28519456
CAS-Nummer: 1783382-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR