Dibromo(fluoro)acetic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,2-Dibromo-2-fluoroethanoic acid
PC2253M-1G
PC2253M-5G
PC2253M-250MG
APOSPC2253M-1GEA
142
EUR
APOSPC2253M-1G
APOSPC2253M-5G
APOSPC2253M-250MG
Dibromo(fluoro)acetic acid 97%
Dibromo(fluoro)acetic acid
Formel:
C₂HBr₂FO₂ Molecular Weight: 235,835 g/mol Siedepunkt: 130 °C (60 mm Hg) Schmelzpunkt: 40…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069082 CAS-Nummer: 353-99-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...