2-Bromo-1,1,1,3,3,3-hexafluoro-2-(2-methoxyphenyl)propane 97%

Lieferant: Apollo Scientific
Gefahr

PC2452-1G PC2452-250MG
APOSPC2452-1GEA 382 EUR
APOSPC2452-1G APOSPC2452-250MG
2-Bromo-1,1,1,3,3,3-hexafluoro-2-(2-methoxyphenyl)propane 97%
2-Bromo-1,1,1,3,3,3-hexafluoro-2-(2-methoxyphenyl)propane
Formel: C₁₀H₇BrF₆O
Molecular Weight: 337,056 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458068
CAS-Nummer: 70430-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR