4-Bromo-5-(trifluoromethoxy)benzene-1,2-diamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-5-(trifluoromethoxy)phenylene-1,2-diamine

PC2478-1G PC2478-5G
APOSPC2478-1GEA 125 EUR
APOSPC2478-1G APOSPC2478-5G
4-Bromo-5-(trifluoromethoxy)benzene-1,2-diamine 97%
4-Bromo-5-(trifluoromethoxy)benzene-1,2-diamine
Formel: C₇H₆BrF₃N₂O
Molecular Weight: 271.035 g/mol
Schmelzpunkt: 39…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07777124
CAS-Nummer: 156425-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR