4-(Pentafluoroethyl)aniline hydrochloride 95%

Lieferant: Apollo Scientific

PC250029-250MG PC250029-500MG PC250029-1G
APOSPC250029-250MGEA 705 EUR
APOSPC250029-250MG APOSPC250029-500MG APOSPC250029-1G
4-(Pentafluoroethyl)aniline hydrochloride 95%
4-(Pentafluoroethyl)aniline hydrochloride
Formel: C₈H₆F₅N·ClH
Molecular Weight: 247,593 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26935948
CAS-Nummer: 60979-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR