1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulphonyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5H-Octafluoro-3-oxapentanesulphonyl fluoride

PC2511-1G PC2511-5G
APOSPC2511-1GEA 236 EUR
APOSPC2511-1G APOSPC2511-5G
1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulphonyl fluoride 95%
1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulphonyl fluoride
Formel: C₄HF₉O₃S
Molecular Weight: 300,1 g/mol
Siedepunkt: 70 °C (400 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD07778617
CAS-Nummer: 104729-49-9
UN: 2920
ADR: 8+3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR