1H,1H-Perfluoro(2,5,8,11,14-pentamethyl-3,6,9,12,15-oxaoctadecan-1-ol) 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H,1H-Pentatriacontafluoro-(2,5,8,11,14-pentamethyl-3,6,9,12,15-oxaoctadecan-1-ol), 2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-Icosafluoro-2,5,8,11,14-pentakis(trifluoromethyl)-3,6,9,12,15-pentaoxaoctadecan-1-ol
PC2651-25G
PC2651-5G
APOSPC2651-25GEA
691
EUR
APOSPC2651-25G
APOSPC2651-5G
1H,1H-Perfluoro(2,5,8,11,14-pentamethyl-3,6,9,12,15-oxaoctadecan-1-ol) 90%
1H,1H-Perfluoro(2,5,8,11,14-pentamethyl-3,6,9,12,15-oxaoctadecan-1-ol)
Formel:
C₁₈H₃F₃₅O₆ Molecular Weight: 980,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460481 CAS-Nummer: 27617-34-1 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...