2-Chloro-4-fluoro-6-nitrotoluene 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-5-fluoro-2-methylnitrobenzene, 1-Chloro-5-fluoro-2-methyl-3-nitrobenzene

PC2785-1G PC2785-250MG PC2785-5G
APOSPC2785-1GEA 145 EUR
APOSPC2785-1G APOSPC2785-250MG APOSPC2785-5G
2-Chloro-4-fluoro-6-nitrotoluene 98%
2-Chloro-4-fluoro-6-nitrotoluene
Formel: C₇H₅ClFNO₂
Molecular Weight: 189,572 g/mol
Schmelzpunkt: 37…39 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094191
CAS-Nummer: 502496-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR