2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-4,4-difluorobutanoic acid

Lieferant: Apollo Scientific

PC27861-100MG PC27861-1G PC27861-250MG
APOSPC27861-100MGEA 561 EUR
APOSPC27861-100MG APOSPC27861-1G APOSPC27861-250MG
2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-4,4-difluorobutanoic acid
2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-4,4-difluorobutanoic acid
Formel: C₁₉H₁₇F₂NO₄
Molecular Weight: 361,339 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738956
CAS-Nummer: 1260640-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR