(2R)-2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-2-(3-fluorophenyl)acetic acid

Lieferant: Apollo Scientific

PC27865-100MG PC27865-250MG
APOSPC27865-100MGEA 1290 EUR
APOSPC27865-100MG APOSPC27865-250MG
(2R)-2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-2-(3-fluorophenyl)acetic acid
(2R)-2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-2-(3-fluorophenyl)acetic acid
Formel: C₂₃H₁₈FNO₄
Molecular Weight: 391,392 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371711
CAS-Nummer: 1260608-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR