Fmoc-α-Methyl-D-4-fluorophenylalanine

Lieferant: Apollo Scientific

PC27887-1G PC27887-250MG
APOSPC27887-1GEA 459 EUR
APOSPC27887-1G APOSPC27887-250MG
Fmoc-α-Methyl-D-4-fluorophenylalanine
Fmoc-α-Methyl-D-4-fluorophenylalanine
Formel: C₂₅H₂₂FNO₄
Molecular Weight: 419.445 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061622
CAS-Nummer: 1217777-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR