2-Amino-6-chloro-4-fluorotoluene 95+%
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-5-fluoro-2-methylaniline
PC2789-250MG
PC2789-1G
PC2789-5G
APOSPC2789-250MGEA
177
EUR
APOSPC2789-250MG
APOSPC2789-1G
APOSPC2789-5G
2-Amino-6-chloro-4-fluorotoluene 95+%
2-Amino-6-chloro-4-fluorotoluene
Formel:
C₇H₇ClFN Molecular Weight: 159,589 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094195 CAS-Nummer: 886761-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...