3-Bromo-2-chloro-6-fluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27904-1G PC27904-250MG
APOSPC27904-1GEA 609 EUR
APOSPC27904-1G APOSPC27904-250MG
3-Bromo-2-chloro-6-fluorobenzyl fluoride 95%
3-Bromo-2-chloro-6-fluorobenzyl fluoride
Formel: C₇H₄BrClF₂
Molecular Weight: 241,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664896
CAS-Nummer: 2244083-98-3
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR