6-Bromo-3-chloro-2-fluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27906-1G PC27906-250MG
APOSPC27906-1GEA 609 EUR
APOSPC27906-1G APOSPC27906-250MG
6-Bromo-3-chloro-2-fluorobenzyl fluoride 95%
6-Bromo-3-chloro-2-fluorobenzyl fluoride
Formel: C₇H₄BrClF₂
Molecular Weight: 241,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664897
CAS-Nummer: 1628524-82-2
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR