3-Bromo-6-chloro-2-fluorobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC27907-1G
PC27907-250MG
APOSPC27907-1GEA
0
EUR
APOSPC27907-1G
APOSPC27907-250MG
3-Bromo-6-chloro-2-fluorobenzyl fluoride 95%
3-Bromo-6-chloro-2-fluorobenzyl fluoride
Formel:
C₇H₄BrClF₂ Molecular Weight: 241,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31664898 CAS-Nummer: 943830-50-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...