2-Bromo-3,6-difluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27911-1G PC27911-250MG
APOSPC27911-1GEA 609 EUR
APOSPC27911-1G APOSPC27911-250MG
2-Bromo-3,6-difluorobenzyl fluoride 95%
2-Bromo-3,6-difluorobenzyl fluoride
Formel: C₇H₄BrF₃
Molecular Weight: 225.006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664900
CAS-Nummer: 2244085-54-7
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR