2-Bromo-3-chloro-6-fluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27913-250MG
APOSPC27913-250MGEA 79.9 EUR
APOSPC27913-250MG
2-Bromo-3-chloro-6-fluorobenzyl fluoride 95%
2-Bromo-3-chloro-6-fluorobenzyl fluoride
Formel: C₇H₄BrClF₂
Molecular Weight: 241.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664901
CAS-Nummer: 2244087-33-8
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR