3-Bromo-4-fluorobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-fluoro-4-(fluoromethyl)benzene
PC27930-1G
PC27930-250MG
APOSPC27930-1GEA
609
EUR
APOSPC27930-1G
APOSPC27930-250MG
3-Bromo-4-fluorobenzyl fluoride 95%
3-Bromo-4-fluorobenzyl fluoride
Formel:
C₇H₅BrF₂ Molecular Weight: 207,015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28527820 CAS-Nummer: 1783551-07-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...