5-Bromo-2-fluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27935-1G PC27935-250MG
APOSPC27935-1GEA 609 EUR
APOSPC27935-1G APOSPC27935-250MG
5-Bromo-2-fluorobenzyl fluoride 95%
5-Bromo-2-fluorobenzyl fluoride
Formel: C₇H₅BrF₂
Molecular Weight: 207,015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664909
CAS-Nummer: 2244085-31-0
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR