2-Amino-5-(trifluoromethyl)benzenesulphonamide 95%

Lieferant: Apollo Scientific
Achtung

PC2825-1G PC2825-250MG
APOSPC2825-1GEA 980 EUR
APOSPC2825-1G APOSPC2825-250MG
2-Amino-5-(trifluoromethyl)benzenesulphonamide 95%
2-Amino-5-(trifluoromethyl)benzenesulphonamide
Formel: C₇H₇F₃N₂O₂S
Molecular Weight: 240.203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741369
CAS-Nummer: 201224-78-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR