Ethyl 7-bromo-5-(trifluoromethoxy)benzofuran-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC28252-1G
PC28252-250MG
APOSPC28252-1GEA
261
EUR
APOSPC28252-1G
APOSPC28252-250MG
Ethyl 7-bromo-5-(trifluoromethoxy)benzofuran-2-carboxylate
Ethyl 7-bromo-5-(trifluoromethoxy)benzofuran-2-carboxylate
Formel:
C₁₂H₈BrF₃O₄ Molecular Weight: 353,089 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31793558 CAS-Nummer: 2379918-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...