4-Bromo-1-(4-chlorophenyl)-4,4-difluorobutane-1,3-dione

Lieferant: Apollo Scientific
Achtung

PC28283-1G PC28283-250MG
APOSPC28283-1GEA 546 EUR
APOSPC28283-1G APOSPC28283-250MG
4-Bromo-1-(4-chlorophenyl)-4,4-difluorobutane-1,3-dione
4-Bromo-1-(4-chlorophenyl)-4,4-difluorobutane-1,3-dione
Formel: C₁₀H₆BrClF₂O₂
Molecular Weight: 311.507 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042701
CAS-Nummer: 1823188-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR