4-Ethoxy-1,1,1-trifluoro-3-methylbut-3-en-2-one

Lieferant: Apollo Scientific
Achtung

PC28305-1G PC28305-250MG PC28305-5G
APOSPC28305-1GEA 685 EUR
APOSPC28305-1G APOSPC28305-250MG APOSPC28305-5G
4-Ethoxy-1,1,1-trifluoro-3-methylbut-3-en-2-one
4-Ethoxy-1,1,1-trifluoro-3-methylbut-3-en-2-one
Formel: C₇H₉F₃O₂
Molecular Weight: 182.14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30730450
CAS-Nummer: 153085-12-2
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR