3-Fluoro-5-methoxyphenylacetonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Cyanomethyl)-5-fluoroanisole, 2-(3-Fluoro-5-methoxyphenyl)acetonitrile, 3-Fluoro-5-methoxybenzyl cyanide

PC2842-1G PC2842-250MG PC2842-5G
APOSPC2842-1GEA 52.2 EUR
APOSPC2842-1G APOSPC2842-250MG APOSPC2842-5G
3-Fluoro-5-methoxyphenylacetonitrile
3-Fluoro-5-methoxyphenylacetonitrile
Formel: C₉H₈FNO
Molecular Weight: 165,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741379
CAS-Nummer: 914637-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR