2-Bromo-6-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-2-carboxybenzotrifluoride, 6-Bromo-α,α,α-trifluoro-o-toluic acid
PC2856-1G
PC2856-5G
PC2856-250MG
APOSPC2856-1GEA
98.9
EUR
APOSPC2856-1G
APOSPC2856-5G
APOSPC2856-250MG
2-Bromo-6-(trifluoromethyl)benzoic acid 97%
2-Bromo-6-(trifluoromethyl)benzoic acid
Formel:
C₈H₄BrF₃O₂ Molecular Weight: 269,015 g/mol Schmelzpunkt: 122…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06253757 CAS-Nummer: 177420-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...