3-Chloro-5-fluoro-4-iodoaniline 95%

Lieferant: Apollo Scientific
Achtung

PC29356-1G PC29356-250MG
APOSPC29356-1GEA 812 EUR
APOSPC29356-1G APOSPC29356-250MG
3-Chloro-5-fluoro-4-iodoaniline 95%
3-Chloro-5-fluoro-4-iodoaniline
Formel: C₆H₄ClFIN
Molecular Weight: 271.459 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32066743
CAS-Nummer: 1849390-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR