7-Bromo-1-fluorophenazine 95%

Lieferant: Apollo Scientific
Achtung

PC29358-1G PC29358-250MG PC29358-5G
APOSPC29358-1GEA 224 EUR
APOSPC29358-1G APOSPC29358-250MG APOSPC29358-5G
7-Bromo-1-fluorophenazine 95%
7-Bromo-1-fluorophenazine
Formel: C₁₂H₆BrFN₂
Molecular Weight: 277.092 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32066753
CAS-Nummer: 2366994-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR