4-Fluoro-2-methylbenzoyl chloride 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Fluoro-o-toluoyl chloride, 2-(Chlorocarbonyl)-5-fluorotoluene

PC2985-1G PC2985-25G PC2985-5G PC2985-100G
APOSPC2985-1GEA 17.6 EUR
APOSPC2985-1G APOSPC2985-25G APOSPC2985-5G APOSPC2985-100G
4-Fluoro-2-methylbenzoyl chloride 99%
4-Fluoro-2-methylbenzoyl chloride
Formel: C₈H₆ClFO
Molecular Weight: 172.584 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115881
CAS-Nummer: 21900-43-6
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR