3-Nitro-2-(2,2,3,3-tetrafluoropropoxy)pyridine

Lieferant: Apollo Scientific

PC300011-1G PC300011-250MG PC300011-500MG PC300011-2.5G
APOSPC300011-1GEA 519 EUR
APOSPC300011-1G APOSPC300011-250MG APOSPC300011-500MG APOSPC300011-2.5G
3-Nitro-2-(2,2,3,3-tetrafluoropropoxy)pyridine
3-Nitro-2-(2,2,3,3-tetrafluoropropoxy)pyridine
Formel: C₈H₆F₄N₂O₃
Molecular Weight: 254,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28967936
CAS-Nummer: 1980049-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR