3-Bromo-6-(trifluoromethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC300500-1G PC300500-100MG
APOSPC300500-1GEA 79.9 EUR
APOSPC300500-1G APOSPC300500-100MG
3-Bromo-6-(trifluoromethyl)-1H-indazole
3-Bromo-6-(trifluoromethyl)-1H-indazole
Formel: C₈H₄BrF₃N₂
Molecular Weight: 265,03 g/mol
Schmelzpunkt: 184…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09263228
CAS-Nummer: 1000341-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR