5-(Trifluoromethyl)-1,3-benzoxazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Trifluoromethyl)benzo[d]oxazole
PC300509-1G
PC300509-250MG
PC300509-5G
APOSPC300509-1GEA
248
EUR
APOSPC300509-1G
APOSPC300509-250MG
APOSPC300509-5G
5-(Trifluoromethyl)-1,3-benzoxazole 95%
5-(Trifluoromethyl)-1,3-benzoxazole
Formel:
C₈H₄F₃NO Molecular Weight: 187,119 g/mol Siedepunkt: 40…42 °C (1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD19374815 CAS-Nummer: 1267217-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...