2-Methyl-5-(trifluoromethyl)-1,3-benzoxazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-5-(trifluoromethyl)benzo[d]oxazole
PC300510-1G
PC300510-250MG
PC300510-5G
APOSPC300510-1GEA
219
EUR
APOSPC300510-1G
APOSPC300510-250MG
APOSPC300510-5G
2-Methyl-5-(trifluoromethyl)-1,3-benzoxazole 97%
2-Methyl-5-(trifluoromethyl)-1,3-benzoxazole
Formel:
C₉H₆F₃NO Schmelzpunkt: 34…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09222481 CAS-Nummer: 175785-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...