(3E)-4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one

Lieferant: Apollo Scientific
Achtung

PC300525-1G PC300525-5G
APOSPC300525-1GEA 176 EUR
APOSPC300525-1G APOSPC300525-5G
(3E)-4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
(3E)-4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
Formel: C₆H₈F₃NO
Molecular Weight: 167,129 g/mol
Schmelzpunkt: 56…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00764476
CAS-Nummer: 127223-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR