Ethyl 3-bromo-5-(trifluoromethoxy)-1H-indole-2-carboxylate

Lieferant: Apollo Scientific

PC300551-1G PC300551-250MG
APOSPC300551-1GEA 290 EUR
APOSPC300551-1G APOSPC300551-250MG
Ethyl 3-bromo-5-(trifluoromethoxy)-1H-indole-2-carboxylate
Ethyl 3-bromo-5-(trifluoromethoxy)-1H-indole-2-carboxylate
Formel: C₁₂H₉BrF₃NO₃
Molecular Weight: 352.104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11615815
CAS-Nummer: 1227955-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR