2-Amino-6-(trifluoromethyl)quinazoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-6-(trifluoromethyl)-1,3-benzodiazine, 6-(Trifluoromethyl)quinazolin-2-amine
PC300554-1G
PC300554-250MG
APOSPC300554-1GEA
221
EUR
APOSPC300554-1G
APOSPC300554-250MG
2-Amino-6-(trifluoromethyl)quinazoline
2-Amino-6-(trifluoromethyl)quinazoline
Formel:
C₉H₆F₃N₃ Molecular Weight: 213,159 g/mol Schmelzpunkt: 184…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00835296 CAS-Nummer: 190273-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...