3-Amino-3-[4-(trifluoromethyl)phenyl]propanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-3-(4-(trifluoromethyl)phenyl)propanoic acid

PC300557-1G PC300557-5G PC300557-250MG
APOSPC300557-1GEA 50.9 EUR
APOSPC300557-1G APOSPC300557-5G APOSPC300557-250MG
3-Amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
3-Amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571869
CAS-Nummer: 180263-44-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR