3-Chloro-2-(methylthio)-5-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC300580-10G
PC300580-1G
PC300580-5G
APOSPC300580-10GEA
625
EUR
APOSPC300580-10G
APOSPC300580-1G
APOSPC300580-5G
3-Chloro-2-(methylthio)-5-(trifluoromethyl)pyridine
3-Chloro-2-(methylthio)-5-(trifluoromethyl)pyridine
Formel:
C₇H₅ClF₃NS Molecular Weight: 227,635 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17168287 CAS-Nummer: 1378308-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...