[4-(3-Fluorophenoxy)phenyl]methanol
Lieferant: Apollo Scientific
PC300614-1G
PC300614-5G
APOSPC300614-1GEA
248
EUR
APOSPC300614-1G
APOSPC300614-5G
[4-(3-Fluorophenoxy)phenyl]methanol
[4-(3-Fluorophenoxy)phenyl]methanol
Formel:
C₁₃H₁₁FO₂ Molecular Weight: 218,224 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11190439 CAS-Nummer: 1039900-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...